Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mov10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163440.1 | NP_001156912.1 | 12 - 13 | 2142 | 85 | |
NM_001163441.1 | NP_001156913.1 | 12 - 13 | 2018 | 85 | ||
NM_008619.2 | NP_032645.2 | 11 - 12 | 2119 | 85 | ||
GenBank mRNA | AK004542.1 | - | 12 - 13 | 1993 | 85 | |
AK150204.1 | - | 12 - 13 | 2002 | 85 | ||
AK159295.1 | - | 12 - 13 | 2153 | 85 | ||
AK169206.1 | - | 12 - 13 | 2113 | 85 | ||
AK173224.1 | - | 11 - 12 | 2032 | 85 | ||
BC053743.1 | - | 11 - 12 | 2038 | 6510959 | 85 | |
X52574.1 | - | 11 - 12 | 2056 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |