Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500007301.mm.1 |
NM_001164593.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500007301.mm.1 |
NM_001164593.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500007301.mm.1 |
TC1500000907.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500007302.mm.1 |
NM_001164593.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500007302.mm.1 |
NM_001164593.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500007302.mm.1 |
TC1500000907.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500007303.mm.1 |
NM_001164593.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500007303.mm.1 |
NM_001164593.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500007303.mm.1 |
TC1500000907.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500007304.mm.1 |
NM_001164593.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500007304.mm.1 |
NM_001164593.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500007304.mm.1 |
TC1500000907.mm.1 |
MTA TC |