Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033633.3 | NP_001028805.2 | 5 - 6 | 1330 | 63 | |
GenBank mRNA | AK147341.1 | - | 5 - 6 | 1331 | 63 | |
BC119586.1 | - | 6 - 7 | 897 | 40058865 | 63 | |
BC119587.1 | - | 5 - 6 | 839 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |