Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028149.1 | NP_082425.1 | 3 - 4 | 415 | 70 | |
GenBank mRNA | AK144786.1 | - | 3 - 4 | 389 | 70 | |
AK161566.1 | - | 3 - 4 | 383 | 70 | ||
AK220232.1 | - | 3 - 4 | 368 | 70 | ||
BB656518.1 | - | 3 - 4 | 417 | 70 | ||
EF649694.1 | - | 3 - 4 | 162 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |