Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026186.4 | NP_080462.2 | 2 - 3 | 364 | 74 | |
GenBank mRNA | AK004983.2 | - | 2 - 3 | 308 | 74 | |
AK017849.1 | - | 2 - 3 | 309 | 74 | ||
AK030628.1 | - | 2 - 3 | 293 | 74 | ||
AK035158.1 | - | 2 - 3 | 293 | 74 | ||
BC021478.1 | - | 2 - 3 | 278 | 74 | ||
BC079553.1 | - | 2 - 3 | 286 | 6856335 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |