Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109626.1 | NP_001103096.1 | 3 - 4 | 2156 | 93 | |
NM_001109628.1 | NP_001103098.1 | 3 - 4 | 2156 | 93 | ||
NM_026952.2 | NP_081228.2 | 3 - 4 | 2156 | 93 | ||
GenBank mRNA | AK049376.1 | - | 3 - 4 | 1802 | 93 | |
AK129237.1 | - | 3 - 4 | 1295 | 93 | ||
AY072295.1 | - | 3 - 4 | 2147 | 93 | ||
BC116645.1 | - | 3 - 4 | 2139 | 40086381 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |