Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021547.3 | NP_067522.1 | 7 - 8 | 842 | 73 | |
GenBank mRNA | AK041538.1 | - | 7 - 8 | 798 | 73 | |
AK150166.1 | - | 7 - 8 | 786 | 73 | ||
AK150254.1 | - | 7 - 8 | 786 | 73 | ||
AK155124.1 | - | 7 - 8 | 781 | 73 | ||
AK160480.1 | - | 7 - 8 | 762 | 73 | ||
AK205267.1 | - | 3 - 4 | 292 | 73 | ||
BC003313.1 | - | 7 - 8 | 759 | 3155179 | 73 | |
BC058727.1 | - | 8 - 9 | 929 | 73 | ||
X82457.1 | - | 7 - 8 | 826 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |