Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207231.1 | NP_997114.1 | 1 - 2 | 357 | 65 | |
GenBank mRNA | AK032519.1 | - | 1 - 2 | 114 | 65 | |
AK154137.1 | - | 1 - 2 | 77 | 65 | ||
AK171194.1 | - | 1 - 2 | 368 | 65 | ||
BC065791.1 | - | 1 - 2 | 357 | 3824354 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |