Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P203691 |
AK138099.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P203691 |
AK154137.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P203691 |
BC065791.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P203691 |
NM_207231.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P203691 |
AK138099.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P203691 |
AK154137.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P203691 |
BC065791.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P203691 |
NM_207231.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P349413 |
AK032519.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P349413 |
AK032519.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
540630 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
924312 |
AK089780.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
924312 |
AK152189.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
924312 |
J03783.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
924312 |
NM_031168.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
924312 |
X06203.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
924312 |
X54542.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
932279 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
374580 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P129492 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P203691 |
AK138099.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P203691 |
AK154137.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P203691 |
BC065791.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P203691 |
NM_207231.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P203691 |
AK138099.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P203691 |
AK154137.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P203691 |
BC065791.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P203691 |
NM_207231.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053841 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053837 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053807 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053786 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053834 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053803 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053814 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053810 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053825 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053852 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053817 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053820 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053836 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053835 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053805 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053828 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053808 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053829 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053783 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053846 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053818 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053812 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053826 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053827 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053822 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053776 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053823 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053844 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053838 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053815 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053781 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053832 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053840 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053802 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053778 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053850 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053830 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053779 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053849 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053789 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28053788 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07457161 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P129492 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P349413 |
AK032519.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P349413 |
AK032519.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P203691 |
AK138099.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P203691 |
AK154137.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P203691 |
BC065791.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P203691 |
NM_207231.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P203691 |
AK138099.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P203691 |
AK154137.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P203691 |
BC065791.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P203691 |
NM_207231.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P135627 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P129492 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09069016 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09069007 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053810 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053830 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053857 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053779 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053855 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053849 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053773 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053755 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053794 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053761 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053835 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053719 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053752 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053781 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053738 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053850 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053814 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053751 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053864 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053815 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053798 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053776 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053711 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053846 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053791 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053825 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053772 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053721 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053717 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053844 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053796 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053826 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053713 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053828 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053763 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053792 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053836 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053823 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053707 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053812 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053760 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053822 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053709 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053840 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053788 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053743 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053808 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053817 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053783 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053759 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053805 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053827 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053769 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053727 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053746 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053863 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053771 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053757 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053716 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053838 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053818 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053819 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053860 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053852 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053762 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053715 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053774 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053710 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053740 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053829 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053859 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053847 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053841 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053728 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053786 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053748 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28053754 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053776 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053819 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09382073 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09515612 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053803 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09121655 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053812 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053825 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053791 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053783 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053859 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09069027 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053844 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09626018 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053789 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053779 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053841 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09621136 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053800 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09444657 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053822 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09258892 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053860 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053857 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09258893 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053840 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053817 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053798 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053788 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09121654 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053796 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053810 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053795 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053805 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053786 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053814 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053864 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053823 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053781 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053846 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09121652 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09069016 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053855 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09382069 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053852 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053849 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09609900 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053807 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053815 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053802 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09258896 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053850 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28053830 |
- |
- |