Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008469.2 | NP_032495.2 | 1 - 2 | 569 | 78 | |
GenBank mRNA | BC057934.1 | - | 1 - 2 | 548 | 5251453 | 78 |
CN456368.1 | - | 1 - 2 | 204 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |