Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161419.1 | NP_001154891.1 | 1 - 2 | 71 | 66 | |
GenBank mRNA | AK008191.1 | - | 1 - 2 | 60 | 66 | |
AK009883.2 | - | 1 - 2 | 60 | 66 | ||
BC003854.1 | - | 1 - 2 | 44 | 3601516 | 66 | |
BY276643.1 | - | 1 - 2 | 72 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |