Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026191.2 | NP_080467.3 | 10 - 11 | 1447 | 123 | |
GenBank mRNA | AK004610.1 | - | 10 - 11 | 1411 | 123 | |
AK010512.1 | - | 10 - 11 | 1427 | 123 | ||
AK029935.1 | - | 10 - 11 | 1418 | 123 | ||
AK031000.1 | - | 10 - 11 | 1393 | 123 | ||
AK075991.1 | - | 4 - 5 | 453 | 123 | ||
AK135592.1 | - | 10 - 11 | 1384 | 123 | ||
BC058276.1 | - | 10 - 11 | 1365 | 6515479 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |