Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27991093 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27991095 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27991090 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27991092 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P494982 |
BP771378.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P494982 |
NM_026191.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P494982 |
BP771378.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P494982 |
NM_026191.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1975420 |
BC058276.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1975420 |
CF749423.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1975420 |
AK031000.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1975420 |
AK075991.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1975420 |
AK010512.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
592994 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
640984 |
AK039957.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
240750 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
549984 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
235962 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
735333 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
796899 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
524933 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
792313 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
608276 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
310127 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
941197 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
636629 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
521707 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
778837 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
280851 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
216505 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
618315 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
448727 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
402210 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
945267 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
728766 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
16433 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
72454 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
4637 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
25218 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
126862 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P112506 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P173597 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1975420 |
BC058276.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1975420 |
CF749423.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1975420 |
AK031000.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1975420 |
AK075991.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1975420 |
AK010512.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P494982 |
BP771378.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P494982 |
NM_026191.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P494982 |
BP771378.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P494982 |
NM_026191.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991102 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991122 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991090 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991095 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991104 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991112 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991096 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991080 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991117 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991120 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991099 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991114 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991106 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991085 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991088 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991115 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991089 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991108 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991116 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991093 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991078 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991084 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991124 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991075 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991103 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991100 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991097 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991125 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27991113 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07429631 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07429549 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02385882 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P173597 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02385857 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P112506 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P442655 |
BC058276.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P442655 |
CF749423.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P442655 |
AK031000.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P442655 |
AK075991.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P442655 |
AK010512.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P115468 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P112506 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P173597 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09095168 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09612375 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09612380 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09095167 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27991095 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27991092 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27991093 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27991090 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991089 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991070 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991023 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990890 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991090 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991103 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990862 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991021 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990888 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990986 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991125 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990945 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990933 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990962 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991122 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991044 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991115 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991060 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990849 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991097 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991014 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990951 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991116 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990847 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990984 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991114 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990882 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990893 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990985 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990897 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991028 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991042 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991043 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990911 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990947 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991008 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991085 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990899 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991072 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990901 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990990 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990938 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990870 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991027 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990936 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990920 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990884 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990859 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991067 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991104 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991113 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991003 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990869 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991102 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991055 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990856 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991019 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990883 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991096 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990858 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990952 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991056 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991046 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991050 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990904 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990993 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991048 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990929 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990971 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990979 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991029 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990874 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990978 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990922 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990887 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991127 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990907 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991124 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991041 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990876 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990927 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990982 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990975 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991128 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990930 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990860 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990925 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990940 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991075 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990850 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990928 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990948 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990997 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991088 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990880 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990931 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991049 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990913 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990921 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991000 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991024 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991112 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991018 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990867 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991062 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990966 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990953 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990873 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991025 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990885 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990969 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991118 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991093 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990909 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27990915 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27991120 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991037 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991078 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991089 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991069 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991112 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991025 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991093 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09612389 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991031 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991072 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991122 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991062 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991063 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991085 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991026 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991125 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991029 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991067 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991033 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09027177 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991036 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991084 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09110606 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991102 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991116 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991034 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991118 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991120 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991124 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991080 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991090 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09095168 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991097 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991028 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991115 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991070 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991113 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991027 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991076 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991106 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991059 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09506578 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991108 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991073 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991082 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991103 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09584116 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991104 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27991060 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09184538 |
- |
- |