Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284394.1 | NP_001271323.1 | 1 - 2 | 166 | 68 | |
NM_011129.2 | NP_035259.1 | 1 - 2 | 166 | 68 | ||
GenBank mRNA | AK135143.1 | - | 1 - 2 | 167 | 68 | |
BC055101.1 | - | 1 - 2 | 166 | 6591016 | 68 | |
X61452.1 | - | 1 - 2 | 96 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |