Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500015424.mm.1 |
NM_001114140.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015424.mm.1 |
NM_001114140.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015424.mm.1 |
NM_013836.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015424.mm.1 |
NM_013836.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015424.mm.1 |
TC1500001958.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500015426.mm.1 |
NM_001114140.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015426.mm.1 |
NM_001114140.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015426.mm.1 |
NM_013836.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015426.mm.1 |
NM_013836.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015426.mm.1 |
TC1500001958.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500015425.mm.1 |
NM_001114140.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015425.mm.1 |
NM_001114140.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015425.mm.1 |
NM_013836.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015425.mm.1 |
NM_013836.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015425.mm.1 |
TC1500001958.mm.1 |
MTA TC |