Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008702.2 | NP_001008702.1 | 12 - 13 | 1793 | 94 | |
NM_001037905.3 | NP_001032994.1 | 12 - 13 | 1867 | 94 | ||
NM_001102400.1 | NP_001095870.1 | 11 - 12 | 1804 | 94 | ||
NM_023118.5 | NP_075607.2 | 13 - 14 | 2521 | 94 | ||
GenBank mRNA | AK154716.1 | - | 13 - 14 | 2505 | 94 | |
AK162744.1 | - | 11 - 12 | 1775 | 94 | ||
AK162851.1 | - | 12 - 13 | 1838 | 94 | ||
AK165810.1 | - | 12 - 13 | 1748 | 94 | ||
AK168375.1 | - | 12 - 13 | 1840 | 94 | ||
BC006588.1 | - | 11 - 12 | 1688 | 3587226 | 94 | |
BC016887.1 | - | 12 - 13 | 1733 | 4242174 | 94 | |
U18869.1 | - | 13 - 14 | 2454 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |