Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500016218.mm.1 |
NR_028448.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016218.mm.1 |
NR_028448.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016218.mm.1 |
NR_028449.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016218.mm.1 |
NR_028449.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016218.mm.1 |
TC1500002037.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016219.mm.1 |
NM_199198.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016219.mm.1 |
NM_199198.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016219.mm.1 |
NR_028447.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016219.mm.1 |
NR_028447.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016219.mm.1 |
NR_028448.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016219.mm.1 |
NR_028448.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016219.mm.1 |
NR_028449.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016219.mm.1 |
NR_028449.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016219.mm.1 |
TC1500002037.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016217.mm.1 |
NR_028448.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016217.mm.1 |
NR_028448.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016217.mm.1 |
NR_028449.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016217.mm.1 |
NR_028449.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016217.mm.1 |
TC1500002037.mm.1 |
MTA TC |