Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amigo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178114.4 | NP_835215.1 | 1 | 695 | 98 | |
GenBank mRNA | AB078880.1 | - | 1 | 353 | 98 | |
AK030848.1 | - | 1 | 662 | 98 | ||
BC095990.1 | - | 1 | 530 | 5358186 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |