Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tshb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009432.2 | NP_033458.2 | 2 - 3 | 106 | 74 | |
GenBank mRNA | J00644.1 | - | 1 - 2 | 39 | 74 | |
M54943.1 | - | 1 - 2 | 39 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |