Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033425.4 | NP_001028597.2 | 1 - 2 | 138 | 93 | |
NM_001289481.1 | NP_001276410.1 | 1 - 2 | 138 | 93 | ||
NM_001289482.1 | NP_001276411.1 | 1 - 2 | 138 | 93 | ||
NM_001289483.1 | NP_001276412.1 | 1 - 2 | 138 | 93 | ||
NM_001289484.1 | NP_001276413.1 | 1 - 2 | 138 | 93 | ||
GenBank mRNA | CF166413.1 | - | 1 - 2 | 54 | 93 | |
DQ323929.1 | - | 1 - 2 | 106 | 93 | ||
DQ323930.1 | - | 1 - 2 | 106 | 93 | ||
DQ323931.1 | - | 1 - 2 | 106 | 93 | ||
EF152499.1 | - | 1 - 2 | 102 | 93 | ||
EF152500.1 | - | 1 - 2 | 102 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |