Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161628.1 | NP_001155100.1 | 13 - 14 | 2302 | 78 | |
NM_001161629.1 | NP_001155101.1 | 11 - 12 | 1790 | 78 | ||
NM_029946.4 | NP_084222.4 | 30 - 31 | 4409 | 78 | ||
GenBank mRNA | AK016516.1 | - | 11 - 12 | 1895 | 78 | |
AK019850.1 | - | 13 - 14 | 2302 | 78 | ||
AK220412.1 | - | 13 - 14 | 2221 | 78 | ||
BC056955.1 | - | 18 - 19 | 2809 | 6812632 | 78 | |
BC065111.1 | - | 30 - 31 | 4409 | 30536704 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |