Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samm50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178614.4 | NP_848729.1 | 7 - 8 | 771 | 79 | |
GenBank mRNA | AK003990.1 | - | 7 - 8 | 727 | 79 | |
AK039949.1 | - | 7 - 8 | 771 | 79 | ||
AK041783.1 | - | 765 | 79 | |||
AK084395.1 | - | 7 - 8 | 765 | 79 | ||
AK144975.1 | - | 7 - 8 | 772 | 79 | ||
AK159740.1 | - | 7 - 8 | 783 | 79 | ||
AK161378.1 | - | 7 - 8 | 761 | 79 | ||
AK167806.1 | - | 7 - 8 | 765 | 79 | ||
AK168510.1 | - | 7 - 8 | 727 | 79 | ||
AK169066.1 | - | 7 - 8 | 761 | 79 | ||
AK169351.1 | - | 7 - 8 | 728 | 79 | ||
AK177148.1 | - | 4 - 5 | 401 | 79 | ||
AK179856.1 | - | 4 - 5 | 358 | 79 | ||
BC119178.1 | - | 7 - 8 | 765 | 8733927 | 79 | |
BC119180.1 | - | 7 - 8 | 765 | 8733929 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |