Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080420.2 | NP_536345.2 | 1 - 2 | 373 | 56 | |
GenBank mRNA | BC016212.1 | - | 1 - 2 | 380 | 4924899 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |