Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080420.2 | NP_536345.2 | 9 - 10 | 1642 | 93 | |
GenBank mRNA | AK018070.1 | - | 1 - 2 | 126 | 93 | |
AK162992.1 | - | 1 - 2 | 90 | 93 | ||
BC016212.1 | - | 9 - 10 | 1649 | 4924899 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |