Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039684.2 | NP_001034773.2 | 12 - 13 | 1129 | 65 | |
GenBank mRNA | AK030982.1 | - | 12 - 13 | 1109 | 65 | |
BC150995.1 | - | 12 - 13 | 1197 | 9087908 | 65 | |
BC158122.1 | - | 12 - 13 | 1289 | 9088195 | 65 | |
DQ177342.1 | - | 12 - 13 | 1129 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |