Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033274.3 | NP_001028446.2 | 11 - 12 | 3535 | 57 | |
GenBank mRNA | AK220233.1 | - | 10 - 11 | 2456 | 57 | |
BC037388.1 | - | 6 - 7 | 862 | 57 | ||
BC049267.1 | - | 4 - 5 | 793 | 57 | ||
BC057182.1 | - | 4 - 5 | 459 | 57 | ||
BC130273.1 | - | 9 - 10 | 1909 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |