Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il7r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008372.4 | NP_032398.3 | 4 - 5 | 675 | 72 | |
GenBank mRNA | AF078906.1 | - | 4 - 5 | 541 | 72 | |
AK040364.1 | - | 4 - 5 | 658 | 72 | ||
AK042264.1 | - | 4 - 5 | 620 | 72 | ||
AK154215.1 | - | 3 - 4 | 489 | 72 | ||
AK154516.1 | - | 4 - 5 | 635 | 72 | ||
BC089571.1 | - | 4 - 5 | 627 | 6758487 | 72 | |
M29697.1 | - | 4 - 5 | 588 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |