Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgfrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011197.3 | NP_035327.2 | 6 - 7 | 2082 | 74 | |
GenBank mRNA | AF152344.1 | - | 6 - 7 | 2082 | 74 | |
AK173172.1 | - | 5 - 6 | 1856 | 74 | ||
BC145713.1 | - | 6 - 7 | 2094 | 40131016 | 74 | |
BC145715.1 | - | 6 - 7 | 2094 | 40131018 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |