Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc48a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026353.4 | NP_080629.1 | 1 - 2 | 253 | 86 | |
GenBank mRNA | AK002729.1 | - | 1 - 2 | 196 | 86 | |
AK007888.1 | - | 1 - 2 | 202 | 86 | ||
AK088485.1 | - | 1 - 2 | 176 | 86 | ||
AK089421.1 | - | 1 - 2 | 254 | 86 | ||
AK138729.1 | - | 1 - 2 | 189 | 86 | ||
AK150552.1 | - | 1 - 2 | 202 | 86 | ||
AK155824.1 | - | 1 - 2 | 240 | 86 | ||
AK159879.1 | - | 1 - 2 | 219 | 86 | ||
AK159892.1 | - | 1 - 2 | 219 | 86 | ||
AK159898.1 | - | 1 - 2 | 172 | 86 | ||
AK168636.1 | - | 1 - 2 | 185 | 86 | ||
AK170393.1 | - | 1 - 2 | 230 | 86 | ||
AK182411.1 | - | 1 - 2 | 93 | 86 | ||
BC022913.1 | - | 1 - 2 | 199 | 4237364 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |