Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc48a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026353.4 | NP_080629.1 | 2 - 3 | 420 | 64 | |
GenBank mRNA | AB041659.1 | - | 2 - 3 | 1618 | 64 | |
AK002729.1 | - | 2 - 3 | 64 | |||
AK007888.1 | - | 2 - 3 | 369 | 64 | ||
AK088485.1 | - | 2 - 3 | 343 | 64 | ||
AK089421.1 | - | 2 - 3 | 421 | 64 | ||
AK150552.1 | - | 2 - 3 | 369 | 64 | ||
AK155824.1 | - | 2 - 3 | 407 | 64 | ||
AK159879.1 | - | 2 - 3 | 386 | 64 | ||
AK159892.1 | - | 2 - 3 | 386 | 64 | ||
AK159898.1 | - | 2 - 3 | 339 | 64 | ||
AK170393.1 | - | 2 - 3 | 397 | 64 | ||
BC022913.1 | - | 2 - 3 | 366 | 4237364 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |