Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163487.1 | NP_001156959.1 | 6 - 7 | 746 | 71 | |
NM_001163488.1 | NP_001156960.1 | 6 - 7 | 682 | 71 | ||
NM_021514.4 | NP_067489.3 | 5 - 6 | 699 | 71 | ||
GenBank mRNA | AF249894.1 | - | 5 - 6 | 639 | 71 | |
AK002711.2 | - | 6 - 7 | 649 | 71 | ||
AK049773.1 | - | 6 - 7 | 654 | 71 | ||
BC005526.1 | - | 5 - 6 | 663 | 3495173 | 71 | |
BC115774.1 | - | 3 - 4 | 454 | 71 | ||
D21865.1 | - | 5 - 6 | 597 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |