Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163487.1 | NP_001156959.1 | 9 - 10 | 995 | 90 | |
NM_001163488.1 | NP_001156960.1 | 9 - 10 | 931 | 90 | ||
NM_021514.4 | NP_067489.3 | 8 - 9 | 948 | 90 | ||
GenBank mRNA | AF249894.1 | - | 8 - 9 | 888 | 90 | |
AK002711.2 | - | 9 - 10 | 898 | 90 | ||
AK049773.1 | - | 9 - 10 | 903 | 90 | ||
BC005526.1 | - | 8 - 9 | 912 | 3495173 | 90 | |
BC115774.1 | - | 6 - 7 | 703 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |