Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgcs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145942.5 | NP_666054.2 | 1 - 2 | 169 | 100 | |
GenBank mRNA | AK031297.1 | - | 1 - 2 | 58 | 100 | |
AK044835.1 | - | 1 - 2 | 59 | 100 | ||
AK136171.1 | - | 1 - 2 | 58 | 100 | ||
BC023851.1 | - | 1 - 2 | 85 | 5347038 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |