Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195003.1 | NP_001181932.1 | 13 - 14 | 1819 | 77 | |
NM_001195004.1 | NP_001181933.1 | 12 - 13 | 1729 | 77 | ||
NM_177619.3 | NP_808287.1 | 11 - 12 | 1552 | 77 | ||
GenBank mRNA | AK147968.1 | - | 12 - 13 | 1546 | 77 | |
AK170389.1 | - | 12 - 13 | 1676 | 77 | ||
BC024092.1 | - | 11 - 12 | 1392 | 77 | ||
BC048904.1 | - | 11 - 12 | 1507 | 6492124 | 77 | |
BC057102.1 | - | 12 - 13 | 1657 | 6401353 | 77 | |
DQ076247.1 | - | 13 - 14 | 1747 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |