Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195003.1 | NP_001181932.1 | 9 - 10 | 1352 | 90 | |
NM_001195004.1 | NP_001181933.1 | 8 - 9 | 1262 | 90 | ||
NM_177619.3 | NP_808287.1 | 7 - 8 | 1085 | 90 | ||
GenBank mRNA | AK147968.1 | - | 8 - 9 | 90 | ||
AK170389.1 | - | 8 - 9 | 1209 | 90 | ||
BC024092.1 | - | 7 - 8 | 925 | 90 | ||
BC048904.1 | - | 7 - 8 | 1040 | 6492124 | 90 | |
BC057102.1 | - | 8 - 9 | 1190 | 6401353 | 90 | |
DQ076247.1 | - | 9 - 10 | 1280 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |