Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009401.2 | NP_033427.1 | 10 - 11 | 1073 | 72 | |
GenBank mRNA | BC130026.1 | - | 10 - 11 | 1064 | 40140833 | 72 |
U25416.1 | - | 10 - 11 | 1073 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |