Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013375.1 | NP_001013393.1 | 5 - 6 | 813 | 72 | |
GenBank mRNA | AK032265.1 | - | 5 - 6 | 757 | 72 | |
AK154637.1 | - | 5 - 6 | 753 | 72 | ||
AK167248.1 | - | 5 - 6 | 812 | 72 | ||
BC119621.1 | - | 5 - 6 | 786 | 40062238 | 72 | |
BC119622.1 | - | 5 - 6 | 786 | 40062245 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |