Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028787.4 | NP_083063.2 | 3 - 4 | 502 | 68 | |
GenBank mRNA | AK076339.1 | - | 3 - 4 | 496 | 68 | |
AK145848.1 | - | 3 - 4 | 503 | 68 | ||
AK169012.1 | - | 3 - 4 | 364 | 68 | ||
AK171951.1 | - | 3 - 4 | 476 | 68 | ||
BC026858.1 | - | 3 - 4 | 361 | 4014064 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |