Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011505.2 | NP_035635.1 | 11 - 12 | 1178 | 74 | |
GenBank mRNA | AF152924.1 | - | 11 - 12 | 1168 | 74 | |
BC032881.1 | - | 11 - 12 | 1132 | 1224878 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |