Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011505.2 | NP_035635.1 | 1 - 2 | 73 | 83 | |
GenBank mRNA | AF152924.1 | - | 1 - 2 | 63 | 83 | |
AK049001.1 | - | 1 - 2 | 42 | 83 | ||
BY720472.1 | - | 1 - 2 | 74 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |