Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itfg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206335.1 | NP_001193264.1 | 9 - 10 | 1498 | 60 | |
NM_207217.4 | NP_997100.1 | 8 - 9 | 1386 | 60 | ||
GenBank mRNA | AK029427.1 | - | 8 - 9 | 1286 | 60 | |
AK158049.1 | - | 1480 | 60 | |||
AK170646.1 | - | 9 - 10 | 1417 | 60 | ||
BC046457.1 | - | 9 - 10 | 1345 | 4222961 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |