Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291025.1 | NP_001277954.1 | 7 - 8 | 1590 | 76 | |
NM_011822.4 | NP_035952.2 | 6 - 7 | 1410 | 76 | ||
GenBank mRNA | AB008895.1 | - | 6 - 7 | 1365 | 76 | |
AK155173.1 | - | 5 - 6 | 1591 | 76 | ||
AK163680.1 | - | 6 - 7 | 1386 | 76 | ||
AK169284.1 | - | 6 - 7 | 1393 | 76 | ||
AK170431.1 | - | 7 - 8 | 1537 | 76 | ||
AK195137.1 | - | 3 - 4 | 339 | 76 | ||
AK197337.1 | - | 2 - 3 | 190 | 76 | ||
BC003917.1 | - | 4 - 5 | 500 | 76 | ||
BC014287.1 | - | 6 - 7 | 1383 | 4193343 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |