Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021793.2 | NP_068565.1 | 11 - 12 | 2026 | 68 | |
GenBank mRNA | AB045293.1 | - | 11 - 12 | 2025 | 68 | |
AK044384.1 | - | 11 - 12 | 2027 | 68 | ||
AK079893.1 | - | 4 - 5 | 1924 | 68 | ||
BC005722.1 | - | 10 - 11 | 1902 | 3595205 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |