Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc46a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026740.2 | NP_081016.2 | 1 - 2 | 312 | 77 | |
GenBank mRNA | AK003278.1 | - | 1 - 2 | 295 | 77 | |
AK170505.1 | - | 1 - 2 | 305 | 77 | ||
AK220201.1 | - | 1 - 2 | 272 | 77 | ||
BC057976.1 | - | 1 - 2 | 297 | 5052851 | 77 | |
CX219335.1 | - | 1 - 2 | 312 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |