Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007666.3 | NP_031692.2 | 8 - 9 | 1648 | 64 | |
GenBank mRNA | BC107008.1 | - | 7 - 8 | 1389 | 40047565 | 64 |
BC107013.1 | - | 7 - 8 | 1389 | 40048308 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |