Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rangap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146174.1 | NP_001139646.1 | 6 - 7 | 860 | 87 | |
NM_011241.4 | NP_035371.4 | 7 - 8 | 908 | 87 | ||
GenBank mRNA | AK042930.1 | - | 6 - 7 | 825 | 87 | |
AK088768.1 | - | 6 - 7 | 782 | 87 | ||
AK129452.1 | - | 6 - 7 | 789 | 87 | ||
AK133905.1 | - | 6 - 7 | 828 | 88 | ||
AK146670.1 | - | 7 - 8 | 868 | 87 | ||
AK159009.1 | - | 6 - 7 | 813 | 87 | ||
AK159525.1 | - | 6 - 7 | 825 | 87 | ||
AK159930.1 | - | 6 - 7 | 816 | 87 | ||
BC014855.1 | - | 6 - 7 | 816 | 3157225 | 87 | |
BC052862.1 | - | 6 - 7 | 783 | 30063509 | 87 | |
BC066213.1 | - | 6 - 7 | 824 | 30466595 | 87 | |
BC071200.1 | - | 7 - 8 | 1024 | 5032610 | 87 | |
U08110.1 | - | 7 - 8 | 891 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |