Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175121.3 | NP_780330.2 | 14 - 15 | 1690 | 87 | |
GenBank mRNA | AK031615.1 | - | 14 - 15 | 1721 | 87 | |
AK033812.1 | - | 2 - 3 | 230 | 87 | ||
AK129342.1 | - | 14 - 15 | 1651 | 87 | ||
AK164568.1 | - | 8 - 9 | 5453 | 87 | ||
AK169378.1 | - | 4 - 5 | 449 | 87 | ||
BC037675.1 | - | 11 - 12 | 1097 | 87 | ||
BC041108.1 | - | 14 - 15 | 1560 | 4038546 | 87 | |
BC048178.1 | - | 14 - 15 | 1690 | 6825801 | 87 | |
BC049271.1 | - | 7 - 8 | 713 | 87 | ||
BC057454.1 | - | 1 - 2 | 71 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |