Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmbim6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171034.1 | NP_001164505.1 | 8 - 9 | 850 | 61 | |
NM_001171035.1 | NP_001164506.1 | 7 - 8 | 706 | 61 | ||
NM_001171036.1 | NP_001164507.1 | 8 - 9 | 671 | 61 | ||
NM_026669.4 | NP_080945.1 | 7 - 8 | 635 | 61 | ||
GenBank mRNA | AK019865.2 | - | 7 - 8 | 597 | 61 | |
AK036782.1 | - | 7 - 8 | 597 | 61 | ||
AK036994.1 | - | 7 - 8 | 597 | 61 | ||
AK049604.1 | - | 7 - 8 | 586 | 61 | ||
AK050299.1 | - | 7 - 8 | 598 | 61 | ||
AK050323.1 | - | 7 - 8 | 586 | 61 | ||
AK088030.1 | - | 7 - 8 | 597 | 61 | ||
AK088683.1 | - | 7 - 8 | 598 | 61 | ||
AK139901.1 | - | 7 - 8 | 598 | 61 | ||
AK159380.1 | - | 7 - 8 | 597 | 61 | ||
AK166935.1 | - | 7 - 8 | 597 | 61 | ||
AK206236.1 | - | 2 - 3 | 102 | 61 | ||
BC005588.1 | - | 7 - 8 | 580 | 3708463 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |