Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2u1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027816.3 | NP_082092.2 | 1 - 2 | 543 | 79 | |
GenBank mRNA | AK018458.1 | - | 1 - 2 | 519 | 79 | |
AK041477.1 | - | 1 - 2 | 544 | 79 | ||
AK142740.1 | - | 1 - 2 | 483 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |