Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lppr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK014763.1 | - | 4 - 5 | 903 | 97 | |
AK032034.1 | - | 4 - 5 | 1269 | 97 | ||
AK043629.1 | - | 4 - 5 | 893 | 97 | ||
AK076176.1 | - | 4 - 5 | 1034 | 97 | ||
AY512657.1 | - | 4 - 5 | 816 | 97 | ||
BC031879.1 | - | 4 - 5 | 1133 | 4527789 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |